Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromobenzyl alcohol 98%

Supplier: AMBEED, INC
Warning

A605696-25G A605696-5G A605696-100G A605696-10G
103810-476EA 242 USD InStock
103810-476 103840-140 76844-306 76844-308 77117-308 77117-310
4-Bromobenzyl alcohol 98%
4-Bromobenzyl alcohol
Precaución: for Research Use Only


Formula: C₇H₇BrO
Boiling Pt: 267.8 °C (760 mm Hg)
Melting Pt: 75…77 °C
Flash Pt: 115.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00004650
CAS Number: 873-75-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty