Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-3-fluorophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A122180-1G A122180-25G A122180-1G A122180-5G
103800-188EA 6 USD InStock
103800-188 77284-862 77312-326 77312-328
4-Chloro-3-fluorophenylboronic acid 98%
4-Chloro-3-fluorophenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₅BClFO₂
Boiling Pt: 302.6 °C (760 mm Hg)
Flash Pt: 136.8 °C
Storage Temperature: Ambient
MDL Number: MFCD01319010
CAS Number: 137504-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty