Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-ethoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A146587-1G A146587-250MG A146587-10G
103803-684EA 24 USD InStock
103803-684 103841-778 77671-072 77881-142
2-Fluoro-4-ethoxyphenylboronic acid 98%
2-Fluoro-4-ethoxyphenylboronic acid
Precaución: Inert atmosphere, 2…8 °C.


Formula: C₈H₁₀BFO₃
MW: 183.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08701765
CAS Number: 279261-82-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty