Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A268813-5G A268813-100G A268813-10G A268813-1G A268813-25G A268813-5G
103852-838EA 8 USD InStock
103852-838 76772-104 76772-106 76772-108 77092-596 77092-598
4-Ethylphenylboronic acid 98%
4-Ethylphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₁₁BO₂
MW: 149.98
Boiling Pt: 285.1 °C (760 mm Hg)
Melting Pt: 150…155 °C
Density: 1.125
Flash Pt: 126.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00859377
CAS Number: 63139-21-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty