Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Pentylphenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A282217-1G A282217-5G A282217-100G A282217-10G A282217-25G A282217-5G
103813-586EA 5 USD InStock
103813-586 103853-538 76802-210 76802-212 76837-234 76837-236
4-Pentylphenylboronic acid 97%
4-Pentylphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₇BO₂
MW: 192.07 g/mol
Boiling Pt: 328 °C (760 mm Hg)
Melting Pt: 82…84 °C
Flash Pt: 152.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00995151
CAS Number: 121219-12-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty