Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[5-(Trifluoromethyl)-2-pyridinyl]methanol 96%

Supplier: AMBEED, INC
Warning

A230974-1G A230974-5G A230974-1G A230974-5G A230974-100MG A230974-250MG
103810-742EA 44 USD InStock
103810-742 103850-488 77089-360 77089-362 77140-060 77140-062
[5-(Trifluoromethyl)-2-pyridinyl]methanol 96%
[5-(Trifluoromethyl)-2-pyridinyl]methanol
Precaución: for Research Use Only


Formula: C₇H₆F₃NO
MW: 177.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04117762
CAS Number: 31181-84-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty