Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Cyano-2-thiopheneboronic acid 97%

Supplier: AMBEED, INC
Warning

A376070-1G A376070-5G A376070-25G A376070-250MG
103826-578EA 18 USD InStock
103826-578 103837-602 76840-906 76989-930
5-Cyano-2-thiopheneboronic acid 97%
5-Cyano-2-thiopheneboronic acid
Precaución: for Research Use Only


Formula: C₅H₄BNO₂S
Boiling Pt: 369.3 °C (760 mm Hg)
Melting Pt: 175 °C
Flash Pt: 177.1 °C
Storage Temperature: Freezer
MDL Number: MFCD02094029
CAS Number: 305832-67-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty