Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoro-2-hydroxybenzeneboronic acid 97%

Supplier: AMBEED, INC
Warning

A144337-1G A144337-5G A144337-250MG A144337-1G A144337-25G A144337-10G
103803-392EA 9 USD InStock
103803-392 103841-470 76811-900 76811-902 77316-712 77316-714 77671-046
5-Fluoro-2-hydroxybenzeneboronic acid 97%
5-Fluoro-2-hydroxybenzeneboronic acid
Precaución: for Research Use Only


Formula: C₆H₆BFO₃
Boiling Pt: 347.1 °C (760 mm Hg)
Melting Pt: 160…166 °C
Flash Pt: 149.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03095357
CAS Number: 259209-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty