Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Methylpyridin-3-yl)methanol 95%

Supplier: AMBEED, INC
Warning

A118323-1G A118323-10G A118323-1G
103799-034EA 19 USD InStock
103799-034 77136-372 77136-374 77311-714
(5-Methylpyridin-3-yl)methanol 95%
(5-Methylpyridin-3-yl)methanol
Precaución: for Research Use Only


Formula: C₇H₉NO
MW: 123.152 g/mol
Boiling Pt: 255.9 °C (760 mm Hg)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD08236816
CAS Number: 102074-19-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty