Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(5-Nitro-1,3-phenylene)dimethanol 95%

Supplier: AMBEED, INC
Warning

A123555-1G A123555-10G A123555-250MG A123555-25G A123555-1G
103800-414EA 42 USD InStock
103800-414 76766-744 76766-746 76766-748 77312-532
(5-Nitro-1,3-phenylene)dimethanol 95%
(5-Nitro-1,3-phenylene)dimethanol
Precaución: for Research Use Only


Formula: C₈H₉NO₄
Boiling Pt: 421.9 °C (760 mm Hg)
Flash Pt: 191.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00007274
CAS Number: 71176-55-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty