Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-2-chloro-3-methoxybenzeneboronic acid 98%

Supplier: AMBEED, INC
Warning

A199890-1G A199890-5G A199890-25G A199890-5G
103817-148EA 12 USD InStock
103817-148 103847-774 77085-832 77085-834
6-Bromo-2-chloro-3-methoxybenzeneboronic acid 98%
6-Bromo-2-chloro-3-methoxybenzeneboronic acid
Precaución: for Research Use Only


Formula: C₇H₇BBrClO₃
Boiling Pt: 403 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09800872
CAS Number: 957062-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty