Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone 97%

Supplier: AMBEED, INC
Warning

A100633-1G A100633-100MG A100633-250MG A100633-1G
103795-548EA 126 USD InStock
103795-548 103799-222 77188-524 77188-526 77282-528
(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone 97%
(6-Chloroimidazo[1,2-b]pyridazin-3-yl)(phenyl)methanone
Precaución: for Research Use Only


Formula: C₁₃H₈ClN₃O
Storage Temperature: Refrigerator
MDL Number: MFCD00108116
CAS Number: 90734-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty