Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-Chloro-2-pyridinyl)methanol 97%

Supplier: AMBEED, INC
Warning

A118289-1G A118289-100G A118289-10G A118289-25G A118289-1G
103799-026EA 13 USD InStock
103799-026 77108-394 77108-396 77108-398 77311-704
(6-Chloro-2-pyridinyl)methanol 97%
(6-Chloro-2-pyridinyl)methanol
Precaución: for Research Use Only


Formula: C₆H₆ClNO
Boiling Pt: 251.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07774110
CAS Number: 33674-97-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty