Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-Methyl-3-pyridyl)methanamine 95%

Supplier: AMBEED, INC
Warning

A116427-1G A116427-100MG A116427-250MG A116427-1G
103798-734EA 71 USD InStock
103798-734 103824-600 76810-212 76810-214 77297-466
(6-Methyl-3-pyridyl)methanamine 95%
(6-Methyl-3-pyridyl)methanamine
Precaución: for Research Use Only


Formula: C₇H₁₀N₂
Boiling Pt: 219.7±25.0 °C (760 mm Hg)
Flash Pt: 107.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06212657
CAS Number: 56622-54-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty