Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(9H-Fluoren-9-yl)methyl hydrazinecarboxylate 98%

Supplier: AMBEED, INC
Warning

A370160-1G A370160-5G A370160-100G A370160-1G A370160-500G
103826-458EA 5 USD InStock
103826-458 103837-472 76840-720 76978-700 77306-808 77672-854
(9H-Fluoren-9-yl)methyl hydrazinecarboxylate 98%
(9H-Fluoren-9-yl)methyl hydrazinecarboxylate
Precaución: For research use only


Formula: C₁₅H₁₄N₂O₂
Boiling Pt: 485.8 °C (760 mm Hg)
Melting Pt: 170 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00239419
CAS Number: 35661-51-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty