Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9-Phenylcarbazole-3-boronic acid 97%

Supplier: AMBEED, INC
Warning

A205204-1G A205204-5G A205204-100G A205204-10G A205204-250MG A205204-1G A205204-25G A205204-5G
103817-614EA 6 USD InStock
103817-614 103848-280 76873-558 76873-560 76873-562 77086-484 77086-486 77086-488
9-Phenylcarbazole-3-boronic acid 97%
9-Phenylcarbazole-3-boronic acid
Precaución: for Research Use Only


Formula: C₁₈H₁₄BNO₂
Boiling Pt: 466 °C (760 mm Hg)
Flash Pt: 235.6 °C
Storage Temperature: Ambient
MDL Number: MFCD12196936
CAS Number: 854952-58-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty