Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: AMBEED, INC
Warning

A562389-5G A562389-1G A562389-5G A562389-100G A562389-10G A562389-250MG A562389-25G
103828-752EA 6 USD InStock
103828-752 103839-886 76843-984 76843-986 77005-192 77005-194 77005-196 77307-814
2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(Bromomethyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₇H₁₄BBrO₂
Boiling Pt: 180.3 °C (760 mm Hg)
Flash Pt: 62.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09271779
CAS Number: 166330-03-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty