Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Chloro(p-tolyl)methylene)dibenzene 98%

Supplier: AMBEED, INC
Warning

A166298-1G A166298-5G A166298-100G A166298-10G A166298-1G A166298-25G A166298-5G
103806-048EA 5 USD InStock
103806-048 103844-308 76690-050 76690-052 77320-556 77320-558 77320-560
(Chloro(p-tolyl)methylene)dibenzene 98%
(Chloro(p-tolyl)methylene)dibenzene
Precaución: for Research Use Only


Formula: C₂₀H₁₇Cl
Melting Pt: 93…95 °C
Flash Pt: 186.5 °C
Storage Temperature: Freezer
MDL Number: MFCD01073758
CAS Number: 23429-44-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty