Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Dhq)2phal 97%

Supplier: AMBEED, INC
Warning

A179334-1G A179334-5G A179334-100G A179334-10G A179334-250MG A179334-25G A179334-1G
103807-420EA 13 USD InStock
103807-420 103845-766 76784-534 76784-536 76784-538 76784-540 77301-608
(Dhq)2phal 97%
(Dhq)2phal
Precaución: for Research Use Only


Formula: C₄₈H₅₄N₆O₄
Storage Temperature: Ambient
MDL Number: MFCD28014549
CAS Number: 140924-50-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty