Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-1-(2-Nitrovinyl)-3-(trifluoromethyl)benzene 97%

Supplier: AMBEED, INC
Danger

A101856-1G A101856-5G A101856-10G A101856-250MG A101856-5G
103796-180EA 58 USD InStock
103796-180 103821-876 76982-194 76982-196 77282-852
(E)-1-(2-Nitrovinyl)-3-(trifluoromethyl)benzene 97%
(E)-1-(2-Nitrovinyl)-3-(trifluoromethyl)benzene
Precaución: for Research Use Only


Formula: C₉H₆F₃NO₂
Boiling Pt: 265.0±35.0 °C (760 mm Hg)
Melting Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00176733
CAS Number: 115665-96-8
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty