Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Bromophenyl)-N-ethylacrylamide 98%

Supplier: AMBEED, INC
Warning

A568032-5G A568032-100MG A568032-250MG
103799-686EA 6 USD InStock
103799-686 76790-328 77669-502 77669-504
3-(3-Bromophenyl)-N-ethylacrylamide 98%
3-(3-Bromophenyl)-N-ethylacrylamide
Precaución: for Research Use Only


Formula: C₁₁H₁₂BrNO
Melting Pt: 89…91 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00075351
CAS Number: 58473-74-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty