Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chlorocinnamaldehyde 97%

Supplier: AMBEED, INC
Warning

A136933-1G A136933-10G A136933-250MG A136933-5G
103802-376EA 6 USD InStock
103802-376 77272-300 77272-302 77315-302
4-Chlorocinnamaldehyde 97%
4-Chlorocinnamaldehyde
Precaución: for Research Use Only


Formula: C₉H₇ClO
Boiling Pt: 290.4 °C (760 mm Hg)
Flash Pt: 134.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00156880
CAS Number: 49678-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty