Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-3-(4-Propoxyphenyl)acrylic acid 97%

Supplier: AMBEED, INC
Warning

A201249-1G A201249-5G A201249-250MG A201249-1G A201249-5G
103817-282EA 14 USD InStock
103817-282 103847-920 77274-850 77302-744 77302-746
(E)-3-(4-Propoxyphenyl)acrylic acid 97%
(E)-3-(4-Propoxyphenyl)acrylic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₄O₃
Boiling Pt: 364.8 °C (760 mm Hg)
Flash Pt: 140.4 °C
Storage Temperature: Ambient
CAS Number: 151539-67-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty