Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-3-Cyclohexylacrylic acid 98%

Supplier: AMBEED, INC
Warning

A271030-1G A271030-5G A271030-250MG A271030-1G
103813-030EA 76 USD InStock
103813-030 103852-938 77113-904 77305-066
(E)-3-Cyclohexylacrylic acid 98%
(E)-3-Cyclohexylacrylic acid
Precaución: for Research Use Only


Formula: C₉H₁₄O₂
MW: 154.21 g/mol
Boiling Pt: 283.2 °C (760 mm Hg)
Melting Pt: 58…59 °C
Flash Pt: 189.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03002774
CAS Number: 56453-86-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty