Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(H-Homocys-OH)₂ 95%

Supplier: AMBEED, INC
Warning

A243976-1G A243976-5G A243976-10G A243976-1G A243976-25G A243976-5G A243976-100G
103811-558EA 5 USD InStock
103811-558 103851-382 76815-486 77304-292 77304-294 77304-296 77672-184
(H-Homocys-OH)₂ 95%
(H-Homocys-OH)₂
Precaución: for Research Use Only


Formula: C₈H₁₆N₂O₄S₂
MW: 268.36 g/mol
Melting Pt: 281…284 °C
Storage Temperature: Ambient
MDL Number: MFCD00020391
CAS Number: 626-72-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty