Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Iodo-2-(trimethylsilyl)acetylene 97%

Supplier: AMBEED, INC
Danger

A537346-1G A537346-5G A537346-250MG
103828-604EA 6 USD InStock
103828-604 103839-726 76843-760 77169-638
1-Iodo-2-(trimethylsilyl)acetylene 97%
1-Iodo-2-(trimethylsilyl)acetylene
Precaución: for Research Use Only


Formula: C₅H₉ISi
Boiling Pt: 156.4 °C (760 mm Hg)
Flash Pt: 48.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00274201
CAS Number: 18163-47-8
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty