Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Iodomethyl)cyclohexane 95%

Supplier: AMBEED, INC
Warning

A248586-1G A248586-5G A248586-1G A248586-5G A248586-100MG A248586-250MG
103811-828EA 86 USD InStock
103811-828 103851-662 77091-010 77091-012 77140-438 77140-440
(Iodomethyl)cyclohexane 95%
(Iodomethyl)cyclohexane
Precaución: for Research Use Only


Formula: C₇H₁₃I
Boiling Pt: 213.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06410739
CAS Number: 5469-33-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty