Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%

Supplier: AMBEED, INC
Warning

A819521-5G A819521-100G A819521-10G A819521-250MG A819521-25G A819521-5G
103841-124EA 18 USD InStock
103841-124 77293-874 77293-876 77293-878 77308-380 77308-382
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl 97%
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Precaución: for Research Use Only


Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 239…242 °C
Flash Pt: 419 °C
Storage Temperature: Ambient
MDL Number: MFCD00010805
CAS Number: 76189-55-4
Merck Index: 13,01223
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty