Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulfonate 97%

Supplier: AMBEED, INC
Warning

A231820-1G A231820-5G A231820-1G A231820-5G
103810-792EA 13 USD InStock
103810-792 103850-546 77089-440 77089-442
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulfonate 97%
(R)-(-)-2,2-Dimethyl-1,3-dioxolan-4-ylmethyl-p-toluenesulfonate
Precaución: for Research Use Only


Formula: C₁₃H₁₈O₅S
MW: 286.34 g/mol
Boiling Pt: 400 °C (760 mm Hg)
Melting Pt: 41…43 °C
Flash Pt: 113 °C
Storage Temperature: Freezer
MDL Number: MFCD00063233
CAS Number: 23788-74-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty