Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-(Piperidin-3-yl)methanol 95%

Supplier: AMBEED, INC
Warning

A105174-1G A105174-5G A105174-1G A105174-5G
103796-852EA 48 USD InStock
103796-852 103822-580 77283-346 77309-454
(R)-(Piperidin-3-yl)methanol 95%
(R)-(Piperidin-3-yl)methanol
Precaución: for Research Use Only


Formula: C₆H₁₃NO
Boiling Pt: 240.4 °C (760 mm Hg)
Flash Pt: 92.3 °C
Storage Temperature: Ambient
MDL Number: MFCD09751036
CAS Number: 37675-20-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty