Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R)-1-(3-Chloro-2-fluorophenyl)ethylamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A362327-1G A362327-5G A362327-100MG A362327-250MG
103826-314EA 98 USD InStock
103826-314 103837-328 76840-538 76840-540 76978-594 76978-596
(1R)-1-(3-Chloro-2-fluorophenyl)ethylamine hydrochloride 95%
(1R)-1-(3-Chloro-2-fluorophenyl)ethylamine hydrochloride
Precaución: For research use only


Formula: C₈H₉ClFN·ClH
Storage Temperature: Ambient
MDL Number: MFCD12758015
CAS Number: 1253792-97-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty