Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-(3-Chlorophenyl)ethanol 95%

Supplier: AMBEED, INC
Warning

A132328-1G A132328-100MG A132328-250MG A132328-5G
103801-722EA 42 USD InStock
103801-722 76823-054 76823-056 77314-302
(R)-1-(3-Chlorophenyl)ethanol 95%
(R)-1-(3-Chlorophenyl)ethanol
Precaución: for Research Use Only


Formula: C₈H₉ClO
Boiling Pt: 230 °C (760 mm Hg)
Flash Pt: 93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09863670
CAS Number: 120121-01-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty