Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-(tert-Butoxycarbonyl)-4-oxopyrrolidine-2-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A135397-1G A135397-100MG A135397-25G A135397-250MG
103802-146EA 26 USD InStock
103802-146 76972-320 77881-070 77881-069
(R)-1-(tert-Butoxycarbonyl)-4-oxopyrrolidine-2-carboxylic acid 97%
(R)-1-(tert-Butoxycarbonyl)-4-oxopyrrolidine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO₅
Boiling Pt: 390.8±42.0 °C (760 mm Hg)
Flash Pt: 190.1 °C
Storage Temperature: Ambient
MDL Number: MFCD06409980
CAS Number: 364077-84-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty