Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-Boc-2-Isopropylpiperazine 98%

Supplier: AMBEED, INC
Warning

A247383-1G A247383-5G A247383-1G A247383-5G A247383-250MG
103811-764EA 95 USD InStock
103811-764 103851-598 77090-932 77090-934 77113-372
(R)-1-Boc-2-Isopropylpiperazine 98%
(R)-1-Boc-2-Isopropylpiperazine
Precaución: for Research Use Only


Formula: C₁₂H₂₄N₂O₂
Boiling Pt: 298.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07772101
CAS Number: 674792-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty