Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-HomoLeu-OH 95%

Supplier: AMBEED, INC
Warning

A187316-1G A187316-5G A187316-1G A187316-100MG A187316-250MG
103816-004EA 41 USD InStock
103816-004 103846-594 77084-282 77111-628 77111-630
Fmoc-D-HomoLeu-OH 95%
Fmoc-D-HomoLeu-OH
Precaución: for Research Use Only


Formula: C₂₂H₂₅NO₄
MW: 367.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270205
CAS Number: 204320-60-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty