Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-Leu-OH·H2O 97%

Supplier: AMBEED, INC
Warning

A109582-100G A109582-10G A109582-25G A109582-5G
103823-330EA 6 USD InStock
103823-330 77270-780 77270-782 77270-784 77283-874
Boc-D-Leu-OH·H2O 97%
Boc-D-Leu-OH·H2O
Precaución: for Research Use Only


Formula: C₁₁H₂₁NO₄·H₂O
MW: 249.3 g/mol
Boiling Pt: 356 °C (760 mm Hg)
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00065583
CAS Number: 200937-17-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty