Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A173692-1G A173692-100MG A173692-250MG A173692-5G
103806-858EA 32 USD InStock
103806-858 103845-158 76690-250 76690-252 77082-426
(R)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid 95%
(R)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₉NO₄
MW: 229.273 g/mol
Melting Pt: 100…104 °C
Flash Pt: 137.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06202402
CAS Number: 101555-60-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty