Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-amino-2,3-dimethylbutanoic acid 97%

Supplier: AMBEED, INC
Warning

A110668-1G A110668-250MG A110668-100MG
103797-704EA 315 USD InStock
103797-704 76796-804 77878-835
(R)-2-amino-2,3-dimethylbutanoic acid 97%
(R)-2-amino-2,3-dimethylbutanoic acid
Precaución: for Research Use Only


Formula: C₆H₁₃NO₂
Boiling Pt: 217.7 °C (760 mm Hg)
Flash Pt: 85.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01318765
CAS Number: 53940-82-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty