Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Aminoheptanoic acid 97%

Supplier: AMBEED, INC
Warning

A415764-1G A415764-5G A415764-100MG A415764-1MG
103827-300EA 40 USD InStock
103827-300 103838-346 77102-074 77102-076
(R)-2-Aminoheptanoic acid 97%
(R)-2-Aminoheptanoic acid
Precaución: for Research Use Only


Formula: C₇H₁₅NO₂
MW: 145.19 g/mol
Boiling Pt: 251.0±23.0 °C (760 mm Hg)
Flash Pt: 105.6 °C
Storage Temperature: Ambient
MDL Number: MFCD08275758
CAS Number: 44902-01-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty