Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-2-Allylglycine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A227453-1G A227453-5G A227453-10G A227453-250MG A227453-25G A227453-5G A227453-1G
103819-322EA 30 USD InStock
103819-322 103850-228 76815-076 76815-078 77089-000 77089-002 77303-798
D-2-Allylglycine hydrochloride 97%
D-2-Allylglycine hydrochloride


Formula: C₅H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06797041
CAS Number: 108412-04-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty