Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Hydroxy-4-phenylbutyric acid 98%

Supplier: AMBEED, INC
Warning

A205079-1G A205079-5G A205079-1G A205079-5G A205079-10G
103817-592EA 14 USD InStock
103817-592 103848-256 77302-934 77302-936 77879-278
(R)-2-Hydroxy-4-phenylbutyric acid 98%
(R)-2-Hydroxy-4-phenylbutyric acid
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Boiling Pt: 356.9 °C (760 mm Hg)
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00192219
CAS Number: 29678-81-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty