Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Methylbutanoic acid 95%

Supplier: AMBEED, INC
Warning

A192670-1G A192670-5G A192670-5G A192670-100MG A192670-250MG
103816-472EA 69 USD InStock
103816-472 103847-082 77084-956 77671-708 77671-710
(R)-2-Methylbutanoic acid 95%
(R)-2-Methylbutanoic acid
Precaución: for Research Use Only


Formula: C₅H₁₀O₂
Boiling Pt: 176…177 °C (760 mm Hg)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD09029093
CAS Number: 32231-50-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty