Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-methylhexanoic acid ≥95%

Supplier: AMBEED, INC
Warning

A156877-1G A156877-250MG A156877-1G
103804-930EA 68 USD InStock
103804-930 103843-134 77162-996 77318-940
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-methylhexanoic acid ≥95%
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-methylhexanoic acid
Precaución: Sealed in dry, at 2…8°C


Formula: C₂₂H₂₅NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD06202394
CAS Number: 212688-54-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty