Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-Amino-3-(2-chlorophenyl)propanoic acid 97%

Supplier: AMBEED, INC
Warning

A417860-1G A417860-250MG
103827-348EA 76 USD InStock
103827-348 76841-992 77115-910
(R)-3-Amino-3-(2-chlorophenyl)propanoic acid 97%
(R)-3-Amino-3-(2-chlorophenyl)propanoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀ClNO₂
Boiling Pt: 336.5±32.0 °C (760 mm Hg)
Melting Pt: 235…239 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04113642
CAS Number: 740794-79-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty