Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-Amino-g-butyrolactone hydrochloride 95%

Supplier: AMBEED, INC
Warning

A183897-1G A183897-5G A183897-10G A183897-250MG A183897-25G A183897-5G
103815-682EA 23 USD InStock
103815-682 103846-244 76974-536 76974-538 77083-854 77083-856
(R)-3-Amino-g-butyrolactone hydrochloride 95%
(R)-3-Amino-g-butyrolactone hydrochloride
Precaución: for Research Use Only


Formula: C₄H₈ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD12545886
CAS Number: 117752-88-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty