Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-Aminopiperidin-2-one hydrochloride ≥95%

Supplier: AMBEED, INC
Warning

A132225-1G A132225-100MG A132225-250MG
103801-704EA 112 USD InStock
103801-704 77871-618 77871-619
(R)-3-Aminopiperidin-2-one hydrochloride ≥95%
(R)-3-Aminopiperidin-2-one hydrochloride
Precaución: Inert atmosphere, room temperature


Formula: C₅H₁₁ClN₂O
MW: 150.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09752974
CAS Number: 406216-02-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty