Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-Methyl-2-phenylbutanoic acid 97%

Supplier: AMBEED, INC
Warning

A118938-1G A118938-1G A118938-5G A118938-250MG
103799-138EA 121 USD InStock
103799-138 77311-804 77871-514 77871-513
(R)-3-Methyl-2-phenylbutanoic acid 97%
(R)-3-Methyl-2-phenylbutanoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
Boiling Pt: 282 °C (760 mm Hg)
Flash Pt: 179.2 °C
Storage Temperature: Ambient
MDL Number: MFCD11505969
CAS Number: 13491-13-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty