Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-4-Amino-3-hydroxybutanoic acid 95%

Supplier: AMBEED, INC
Warning

A116967-1G A116967-5G A116967-100MG A116967-250MG A116967-1G A116967-5G
103798-808EA 175 USD InStock
103798-808 103824-680 76822-146 76822-148 77311-470 77311-472
(R)-4-Amino-3-hydroxybutanoic acid 95%
(R)-4-Amino-3-hydroxybutanoic acid
Precaución: for Research Use Only


Formula: C₄H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD02262047
CAS Number: 7013-07-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty