Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Methyl 2-(pyrrolidin-2-yl)acetate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A195583-1G A195583-5G A195583-100MG A195583-250MG
103816-766EA 242 USD InStock
103816-766 103847-378 76986-270 76986-272
(R)-Methyl 2-(pyrrolidin-2-yl)acetate hydrochloride 95%
(R)-Methyl 2-(pyrrolidin-2-yl)acetate hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₄ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD22381965
CAS Number: 340040-67-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty