Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Methyl 2-oxooxazolidine-4-carboxylate 97%

Supplier: AMBEED, INC
Warning

A291143-1G A291143-5G A291143-250MG A291143-1G
103813-890EA 21 USD InStock
103813-890 103835-032 76837-618 76988-662 77305-592
(R)-Methyl 2-oxooxazolidine-4-carboxylate 97%
(R)-Methyl 2-oxooxazolidine-4-carboxylate
Precaución: for Research Use Only


Formula: C₅H₇NO₄
Boiling Pt: 354.3 °C (760 mm Hg)
Melting Pt: 160…165 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11505659
CAS Number: 144542-43-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty